Learn More
Bioworld L-Asparagine, 500 g
Supplier:  Bioworld 401002802
Encompass_Preferred
Synonym: (S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide Technical Information:AnhydrousAppearance: White Crystalline PowderDensity : 1.543 g/cm3HS Code : 29241900IUPAC_Name: AsparagineMelting Point: 235 °C
Catalog No. 50-255-287
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.