Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Asparagine, Monohydrate, FCC, 98-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603723 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-723 25 g Amber Glass Bottle
18-603-724 100 g Amber Glass Bottle
18-603-725 1 kg Amber Glass Bottle
18-603-726 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-723 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A137325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

A1373, 5794-13-8, C4H8N2O3·H2O

Spectrum™ Chemical L-Asparagine, Monohydrate, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.13
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
IUPAC Name (2S)-2-amino-3-carbamoylpropanoic acid hydrate
SMILES O.N[C@@H](CC(N)=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 11.5-12.5%
Packaging Amber Glass Bottle
Quantity 25 g
Optical Rotation +33.0° to +36.5°
Percent Purity 98 to 101.5%
Grade FCC
Physical Form Crystals or Flakes
Chemical Name or Material L-Asparagine, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.