Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%

Catalog No. AAH63257MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63257MD 250 mg
AAH6325703 1 g
AAH6325706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63257MD Supplier Thermo Scientific Chemicals Supplier No. H63257MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 2673-19-0 | C9H18ClNO4 | 239.696 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2673-19-0
Molecular Formula C9H18ClNO4
Molecular Weight (g/mol) 239.696
MDL Number MFCD00153427
InChI Key SFYKWYAIJZEDNG-RGMNGODLSA-N
Synonym h-asp otbu-ome.hcl, h-asp otbu-ome hcl, l-aspartic acid 4-tert-butyl-1-methyl ester hydrochloride, h-asp otbu-ome hydrochloride, h-asp obut-ome hcl, s-4-tert-butyl 1-methyl 2-aminosuccinate hydrochloride, 4-tert-butyl 1-methyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-ome, h-asp otbu-omecl, h-asp o-tbu-ome
PubChem CID 13970777
IUPAC Name 4-O-tert-butyl 1-O-methyl (2S)-2-aminobutanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CC(C(=O)OC)N.Cl

Specifications

Beilstein 6005591
Quantity 250 mg
Formula Weight 239.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.