Learn More
Thermo Scientific Chemicals L-Aspartic acid, 98+%
Description
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
Solubility
Soluble in water(4.5g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Melting Point | >300°C (decomposition) |
| Beilstein | 1723530 |
| Merck Index | 14,840 |
| Solubility Information | Soluble in water(4.5g/L). |
| Density | 1.66 |
| IUPAC Name | (2S)-2-aminobutanedioic acid |
| Quantity | 5000 g |
| Odor | Odorless |
| Formula Weight | 133.1 |
| Optical Rotation | +25° (c=2 in 5N HCl) |
| Show More |
Safety and Handling
| EINECSNumber | 200-291-6 |
| RTECSNumber | CI9098500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.