Learn More
Bioworld L-ASPARTIC ACID SEPAROPORE
Supplier:  Bioworld 201810673
Encompass
L-ASPARTIC ACID SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: L-Aspartic Acid; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads); pH stability: 4-9; Flow ...
Catalog No. 50-197-839
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.