Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartyl-L-Phenylalanine methyl ester, 99.8%, MP Biomedicals™

Catalog No. ICN19000701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19000701 1 g
ICN19000705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19000701 Supplier MP Biomedicals, Inc Supplier No. 0219000701
Add to Cart
Add to Cart

Used as a synthetic sweeter, sugar substitute

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
Synonym aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
PubChem CID 134601
ChEBI CHEBI:2877
IUPAC Name (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 294.31
Physical Form Powder
Chemical Name or Material L-Aspartyl-L-Phenylalanine methyl ester
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.