Learn More
Medchemexpress LLC L-azetidine-2-carboxylic acid | 2133-34-8 | 99.9% | 101.10 g/mol | C4H7NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW050044100MG
Encompass_Preferred
L-azetidine-2-carboxylic acid is a non-proteinogenic amino acid and a proline analog used as a research reagent in biochemical and cell biology studies. It is supplied as a high-purity solid for experiments involving protein synthesis, folding, and proline metabolism.
  • Non-proteinogenic amino acid and proline analog.
  • High purity (99.85%) suitable for research applications.
  • Molecular formula C4H7NO2 and molecular weight 101.10 g/mol.
  • Available as a 100 MG solid and in larger solid quantities.
  • Used in studies of protein synthesis, folding, and metabolism.
  • SDS, COA, and data sheet available for handling and quality information.
Catalog No. 50-002-10568