Learn More
Medchemexpress LLC L-azetidine-2-carboxylic acid | 2133-34-8 | 99.9% | 101.10 g/mol | C4H7NO2 | 1 ML

Supplier: Medchemexpress LLC HYW05004410MM/1ML
L-azetidine-2-carboxylic acid is a proline analog research reagent used to study protein folding, endoplasmic reticulum (ER) stress, autophagy, and apoptosis pathways. It is supplied as a solid and as a 10 mM solution in water and has reported high chemical purity.
- Proline analog that perturbs protein synthesis and folding
- Induces ER stress and protein misfolding
- Upregulates LC3-II via PERK activation (autophagy marker)
- Increases pro-apoptotic BAX and induces ATF6 cleavage
- Available as solid and as a 10 mM, 1 mL aqueous solution
- High purity, reported at 99.85%
- CAS number 2133-34-8; formula C4H7NO2; molecular weight 101.10 g/mol
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.