Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L.b. Cas12a Nuclease

Catalog No. A40001795
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 pmol
2000 pmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40001795 2000 pmol
A40001796 200 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A40001795 Supplier Invitrogen™ Supplier No. A40001795
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L.b. Cas12a Nuclease for simple, quick and specific analysis of your samples.

L. b. (Lachnospiraceae bacterium) Cas12a Nuclease, also known as Cpf1, is an RNA-guided endonuclease derived from the Class II type V CRISPR-Cas system. It exhibits a high degree of specificity in targeting and cleaving particular DNA sequences in genome editing. Additionally, Cas12a nuclease possesses collateral cleavage activity, resulting in the non-specific cleavage of neighboring DNA upon target recognition.

The DNA cleavage capability of Cas12a nuclease has been leveraged for various applications, including genome editing, gene regulation, and nucleic acid detection. By programming Cas12a with specific guide RNAs, targeted modifications such as gene knockout, gene insertion, or point mutations can be introduced into the target DNA sequence.

Disclaimer: The use of this product for certain applications may require a license under third party intellectual property rights and it is buyer's sole responsibility to obtain any required license for such use. Thermo Fisher Scientific makes no representation or warranty that the use of this product is free from third party intellectual property rights, and will not indemnify the buyer against any third party infringement claim arising from buyer's use of the product.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 μM
Quantity 2000 pmol
Product Type Stand-alone enzyme
Research Category Nucleic Acid Analysis
Form Liquid

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.