Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-BMAA hydrochloride, Tocris Bioscience™

Catalog No. 253810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-381-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-381-0 Supplier R&D Systems Supplier No. 2538/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Neurotoxic amino acid

Neurotoxic glutamate agonist originally isolated from Cycas circinalis. Implicated in the pathogenesis of amyotrophic lateral sclerosis-Parkinsonism-dementia complex of Guam (Guam ALS-PD).

Chemical Identifiers

CAS 16012-55-8
Molecular Formula C4H11ClN2O2
Molecular Weight (g/mol) 154.594
InChI Key VDXYGASOGLSIDM-DFWYDOINSA-N
Synonym l-bmaa hydrochloride, s-2-amino-3-methylamino propanoic acid hydrochloride, beta-n-methylamino-l-alanine hydrochloride, s +-2-amino-3-methylamino propionic acid hydrochloride, s-2-amino-3-methylamino-propionic acid hydrochloride, l-2-amino-3 methylamino propanoicacidhydrochloride, h-l-dap me-oh hcl, s +-2-amino-3-methylamino propionicacidhydrochloride, bestipharma 572-892, beta-methylamino-l-alanine hcl
PubChem CID 51358333
IUPAC Name (2S)-2-amino-3-(methylamino)propanoic acid;hydrochloride
SMILES CNCC(C(=O)O)N.Cl

Specifications

Quantity 10 mg
Formula Weight 154.6
Percent Purity >98%
Chemical Name or Material L-BMAA hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.