Learn More
United States Biological Corporation L BROTH 'LURIA' BROTH 500G
Catalog No. NC9049544 Shop All United States Biological Corporation Products
Encompass_Preferred
L-Broth (Luria Broth) is the standard nutrient media for propagation of Escherichia coli for purposes of strain maintenance, cloning, plasmid propagation, and protein expression. LB Broth media formulations have been industry standards for the cultivation of Escherichia coli since the 1950s (1,2,3). �These media have been widely used in molecular microbiology applications for the preparation of plasmid DNA and recombinant proteins (4,5). �The media are nutrient-rich formulations which provide peptides and peptones, vitamins, and trace elements. �The three formulations (Lennox, Miller and Luria) differ in the amount of sodium chloride, thus providing selection of the appropriate osmotic conditions for the particular bacterial strain and desired culture conditions. The low salt formulations, Lennox and Luria, are ideal for cultures requiring salt-sensitive antibiotics such as Zeocin.
Catalog No. NC9049544 Supplier United States Biological Corporation Supplier No. L1530500G
L-Broth (Luria Broth) is the standard nutrient media for propagation of Escherichia coli for purposes of strain maintenance, cloning, plasmid propagation, and protein expression. LB Broth media formulations have been industry standards for the cultivation of Escherichia coli since the 1950s (1,2,3). �These media have been widely used in molecular microbiology applications for the preparation of plasmid DNA and recombinant proteins (4,5). �The media are nutrient-rich formulations which provide peptides and peptones, vitamins, and trace elements. �The three formulations (Lennox, Miller and Luria) differ in the amount of sodium chloride, thus providing selection of the appropriate osmotic conditions for the particular bacterial strain and desired culture conditions. The low salt formulations, Lennox and Luria, are ideal for cultures requiring salt-sensitive antibiotics such as Zeocin.