Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Carnitine hydrochloride, 98%

Catalog No. ac230281000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC230281000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC230281000 Supplier Thermo Scientific Chemicals Supplier No. 230281000
Only null left
Add to Cart

CAS: 6645-46-1 | C7H16ClNO3 | 197.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6645-46-1
Molecular Formula C7H16ClNO3
Molecular Weight (g/mol) 197.66
MDL Number MFCD00066100
InChI Key JXXCENBLGFBQJM-FYZOBXCZSA-N
Synonym r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, +--carnitine chloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride
PubChem CID 656657
IUPAC Name [(2R)-3-carboxy-2-hydroxypropyl]-trimethylazanium;chloride
SMILES [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O

Specifications

Melting Point 142.0°C
Loss on Drying 5% max.
Infrared Spectrum Authentic
Linear Formula HO2CCH2CH(OH)CH2N(CH3)3Cl
Quantity 100 g
Merck Index 15,1849
Specific Rotation Condition −22.00 (20.00°C c=1, H2O)
Specific Rotation -22
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot ethanol, slightly soluble in cold ethanol, practically insoluble in acetone and ether
Formula Weight 197.66
Percent Purity 98%
Chemical Name or Material L(-)-Carnitine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 229-663-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.