Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-carnitine hydrochloride, 99%, MP Biomedicals™

Catalog No. ICN19503101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19503101 1 g
ICN19503105 5 g
ICN19503125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19503101 Supplier MP Biomedicals, Inc Supplier No. 0219503101
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6645-46-1
Molecular Formula C7H16ClNO3
Molecular Weight (g/mol) 197.66
MDL Number MFCD00066100
InChI Key JXXCENBLGFBQJM-FYZOBXCZSA-N
Synonym r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, +--carnitine chloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride
PubChem CID 656657
IUPAC Name [(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 197.7
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material L-carnitine hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.