Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteic acid, 99%

Catalog No. AC226120250
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226120250 25 g
AC226120100 10 g
AC226120500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226120250 Supplier Thermo Scientific Chemicals Supplier No. 226120250

CAS: 498-40-8 | C3H7NO5S | 169.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-40-8
Molecular Formula C3H7NO5S
Molecular Weight (g/mol) 169.15
MDL Number MFCD00007524
InChI Key XVOYSCVBGLVSOL-UHFFFAOYNA-N
Synonym l-cysteic acid, cysteinesulfonic acid, l-alanine, 3-sulfo, 3-sulfo-l-alanine, cysteic acid, l, r-2-amino-3-sulfopropanoic acid, cysteic acid van, l-cysteate, cysteinic acid, cysteric acid
PubChem CID 72886
ChEBI CHEBI:17285
SMILES NC(CS(O)(=O)=O)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula HO3SCH2CH(NH2)CO2H
Quantity 25 g
Specific Rotation +7° to +9° (20°C, 589nm) (c=5, H2O)
IUPAC Name 2-amino-3-sulfopropanoic acid
Formula Weight 169.16
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Powder
Chemical Name or Material L-Cysteic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.