Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteic acid monohydrate, 99%, Thermo Scientific Chemicals

Catalog No. AC452840500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452840500 50 g
AC452840100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452840500 Supplier Thermo Scientific Chemicals Supplier No. 452840500

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23537-25-9
Molecular Formula C3H6NO5S
Molecular Weight (g/mol) 168.14
MDL Number MFCD00149544
InChI Key XVOYSCVBGLVSOL-REOHCLBHSA-M
Synonym l-cysteic acid monohydrate, unii-vn8x20t18r, cysteic acid monohydrate, l, r-2-amino-3-sulfopropanoic acid hydrate, l-alanine, 3-sulfo-, monohydrate, cysteinesulfonic acid hydrate, h-cys o 2-oh.h2o, l-cysteic acid hydrate, l-cysteic acid monohydrate mi, alanine, 3-sulfo-, monohydrate, l
PubChem CID 12308854
IUPAC Name (2R)-2-amino-3-sulfopropanoic acid;hydrate
SMILES [NH3+][C@@H](CS([O-])(=O)=O)C([O-])=O

Specifications

Melting Point 253.0°C to 254.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Linear Formula HO3SCH2CH(NH2)CO2H·H2O
Quantity 50 g
Specific Rotation Condition + 8.00 (20.00°C c=5, H2O)
Specific Rotation + 8.00
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 187.17
Percent Purity 99%
Chemical Name or Material L-Cysteic acid monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.