Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18611207 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 kg
50 kg
Packaging:
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-207 25 kg Fiber Drum
18-611-208 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-611-207 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CY11025KGBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 52-90-4

Specifications

Ammonium 0.0002
Arsenic (As) 2e-06
CAS Max % 1
Chloride 0.0005
Heavy Metals 1e-05
Iron (Fe) 3e-05
Loss on Drying 0.005
Packaging Fiber Drum
Residue after Ignition 0.002
Specific Rotation +7.0° to +9.0°
Sulfate 0.0003
Transmittance 0.95
Quantity 25 kg
Percent Purity 98 to 102%
Grade Reagent
Chemical Name or Material L-Cysteine
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.