CAS RN 52-90-4
Thermo Scientific Chemicals L-Cysteine, 98+%
CAS: 52-90-4 Molecular Formula: C3H7NO2S Molecular Weight (g/mol): 121.154 MDL Number: MFCD00064306 InChI Key: XUJNEKJLAYXESH-REOHCLBHSA-N Synonym: l-cysteine,cysteine,cystein,half-cystine,r-cysteine,thioserine,r-2-amino-3-mercaptopropanoic acid,l-+-cysteine,l-cystein,2r-2-amino-3-sulfanylpropanoic acid PubChem CID: 5862 ChEBI: CHEBI:17561 IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid SMILES: C(C(C(=O)O)N)S
L-Cysteine, 99.5%, MP Biomedicals™
CAS: 52-90-4 Molecular Formula: C3H7NO2S Molecular Weight (g/mol): 121.154 InChI Key: XUJNEKJLAYXESH-REOHCLBHSA-N Synonym: l-cysteine,cysteine,cystein,half-cystine,r-cysteine,thioserine,r-2-amino-3-mercaptopropanoic acid,l-+-cysteine,l-cystein,2r-2-amino-3-sulfanylpropanoic acid PubChem CID: 5862 ChEBI: CHEBI:17561 IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid SMILES: C(C(C(=O)O)N)S
L-Cysteine , MP Biomedicals, LLC
CAS: 52-90-4 Molecular Formula: C3H7NO2S Molecular Weight (g/mol): 121.15 InChI Key: XUJNEKJLAYXESH-UHFFFAOYNA-N Synonym: l-cysteine,cysteine,cystein,half-cystine,r-cysteine,thioserine,r-2-amino-3-mercaptopropanoic acid,l-+-cysteine,l-cystein,2r-2-amino-3-sulfanylpropanoic acid PubChem CID: 5862 ChEBI: CHEBI:17561
L-Cysteine [Free Base], MP Biomedicals
CAS: 52-90-4 Molecular Formula: C3H7NO2S Molecular Weight (g/mol): 121.154 InChI Key: XUJNEKJLAYXESH-REOHCLBHSA-N Synonym: l-cysteine,cysteine,cystein,half-cystine,r-cysteine,thioserine,r-2-amino-3-mercaptopropanoic acid,l-+-cysteine,l-cystein,2r-2-amino-3-sulfanylpropanoic acid PubChem CID: 5862 ChEBI: CHEBI:17561 IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid SMILES: C(C(C(=O)O)N)S
MilliporeSigma™ L-Cysteine, Free Base, ≥98%, Calbiochem™,
Format: Glass Bottle
Thermo Scientific Chemicals L-Cysteine, Cell Culture Reagent
CAS: 52-90-4 Molecular Formula: C3H7NO2S Molecular Weight (g/mol): 121.154 MDL Number: MFCD00064306 InChI Key: XUJNEKJLAYXESH-REOHCLBHSA-N Synonym: l-cysteine,cysteine,cystein,half-cystine,r-cysteine,thioserine,r-2-amino-3-mercaptopropanoic acid,l-+-cysteine,l-cystein,2r-2-amino-3-sulfanylpropanoic acid PubChem CID: 5862 ChEBI: CHEBI:17561 IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid SMILES: C(C(C(=O)O)N)S
L-Cysteine, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Sigma Aldrich Azidobenzene Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Thermo Scientific Production Chemicals and Services L-Cysteine DAB, Ajinomoto
CAS: 52-90-4 Molecular Weight (g/mol): 121.16
Sigma Aldrich L-Cysteine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Thermo Scientific Production Chemicals and Services L-Cysteine DAB, Ajinomoto
CAS: 52-90-4 Molecular Weight (g/mol): 121.16
Thermo Scientific Production Chemicals and Services L-Cysteine DAB, Ajinomoto
CAS: 52-90-4 Molecular Weight (g/mol): 121.16
Sigma Aldrich L-Cysteine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2