Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine DAB, Ajinomoto

Catalog No. 502946023
Encompass
Change view
Click to view available options
20 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6023 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6023 Supplier Thermo Scientific Production Chemicals and Services Supplier No. A21313
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 52-90-4
Molecular Weight (g/mol) 121.16

Specifications

Packaging Fiber Box
Quantity 20 kg
Chemical Name or Material L-Cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.