Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine [Free Base], MP Biomedicals

Catalog No. ICN19464691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19464691 1 kg
ICN19464625 25 g
ICN19464680 100 g
ICN19464690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19464691 Supplier MP Biomedicals, Inc Supplier No. 0219464691
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 52-90-4 | C3H7NO2S | 121.154 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Quantity 1 kg
Formula Weight 121.2
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Cysteine [Free Base]
Recommended Storage Room temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.