Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine, Spectrum™ Chemical
SDP

Catalog No. 18621713 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
10 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-713 10 kg
18-621-709 25 g
18-621-710 100 g
18-621-711 500 g
18-621-712 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-621-713 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CY11010KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

CAS: 52-90-4 | 121.16 g/mol

Chemical Identifiers

CAS 52-90-4

Specifications

Packaging Fiber Drum
Molecular Weight (g/mol) 121.16
Quantity 10 kg
Percent Purity 98 to 102%
Grade Reagent
Chemical Name or Material L-Cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.