Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine, 99%

Catalog No. AC111805000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111805000 500 g Plastic Bottle
AC111801000 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111805000 Supplier Thermo Scientific Chemicals Supplier No. 111805000

L-Cystine, CAS # 56-89-3 is the left-handed enantiomer of the sulphur-containing amino acid, cystine, purified for use in molecular biology and analytical chemistry applications. | CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • L-Cystine is a derivative of the amino acid, cysteine
  • Cystine is not considered one of the twenty common amino acids, but it has a role as a metabolite in some species
  • May act as an antioxidant in form and reduce aging, inflammation, and tissue damage

Applications

  • Used as an additive to cell culture media
  • Intended for biochemistry applications such as HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
MDL Number MFCD00064228
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
PubChem CID 67678
ChEBI CHEBI:16283
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Ammonium 200ppm max.
Color White to Beige
Chloride 200ppm max.
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 0.5 % max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula [-SCH2CH(NH2)CO2H]2
Beilstein 04, 507
Specific Rotation −209° to −224° (20°C, 589nm) (c=2, 1 M HCl) on dried substance
Sulfate 300ppm max.
Merck Index 15, 2779
Sulfated Ash 0.1 % max.
Specific Rotation Condition − 224.00 (20.00°C c=2;1 M HCL dry.sub.)
Quantity 500 g
Formula Weight 240.3
Percent Purity ≥98.5% (Acid)
Physical Form Fused Solid, Crystals or Chunks
Chemical Name or Material L-Cystine
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.