CAS RN 56-89-3
L-Cystine, 99%
CAS: 56-89-3 Molecular Formula: C6H12N2O4S2 Molecular Weight (g/mol): 240.292 MDL Number: MFCD00064228 InChI Key: LEVWYRKDKASIDU-IMJSIDKUSA-N Synonym: l-cystine,cystine,l-dicysteine,l-cystin,cystine acid,cysteine disulfide,oxidized l-cysteine,beta,beta'-dithiodialanine,h-cys-oh 2,l-cysteine disulfide PubChem CID: 67678 ChEBI: CHEBI:16283 IUPAC Name: (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid SMILES: C(C(C(=O)O)N)SSCC(C(=O)O)N
L-Cystine, Cell Culture Reagent
CAS: 56-89-3 Molecular Formula: C6H12N2O4S2 Molecular Weight (g/mol): 240.292 MDL Number: MFCD00064228 InChI Key: LEVWYRKDKASIDU-IMJSIDKUSA-N Synonym: l-cystine,cystine,l-dicysteine,l-cystin,cystine acid,cysteine disulfide,oxidized l-cysteine,beta,beta'-dithiodialanine,h-cys-oh 2,l-cysteine disulfide PubChem CID: 67678 ChEBI: CHEBI:16283 IUPAC Name: (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid SMILES: C(C(C(=O)O)N)SSCC(C(=O)O)N
L-Cystine , MP Biomedicals, LLC
CAS: 56-89-3 Molecular Formula: C6H12N2O4S2 Molecular Weight (g/mol): 240.29 InChI Key: LEVWYRKDKASIDU-IMJSIDKUSA-N Synonym: l-cystine,cystine,l-dicysteine,l-cystin,cystine acid,cysteine disulfide,oxidized l-cysteine,beta,beta'-dithiodialanine,l-cysteine disulfide,--cystine PubChem CID: 67678 ChEBI: CHEBI:16283
L-Cystine, FCC, 98.5-101.5%, Spectrum™ Chemical
CAS: 56-89-3 Molecular Formula: C6H12N2O4S2 Molecular Weight (g/mol): 240.29 InChI Key: LEVWYRKDKASIDU-IMJSIDKUSA-N IUPAC Name: (2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid SMILES: N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
L-Cystine, 99%, MP Biomedicals™
CAS: 56-89-3 Molecular Formula: C6H12N2O4S2 Molecular Weight (g/mol): 240.292 MDL Number: MFCD00064228 InChI Key: LEVWYRKDKASIDU-IMJSIDKUSA-N Synonym: l-cystine,cystine,l-dicysteine,l-cystin,cystine acid,cysteine disulfide,oxidized l-cysteine,beta,beta'-dithiodialanine,h-cys-oh 2,l-cysteine disulfide PubChem CID: 67678 ChEBI: CHEBI:16283 IUPAC Name: (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid SMILES: C(C(C(=O)O)N)SSCC(C(=O)O)N
Sigma Aldrich L-Cystine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Thermo Scientific Production Chemicals and Services L-Cystine EP/FCC, Ajinomoto
CAS: 56-89-3 Molecular Weight (g/mol): 240.3
Sigma Aldrich Gamma-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MilliporeSigma™ L-Cystine, ≥98%, Calbiochem™,
CAS: 56-89-3 Molecular Formula: C6H12N2O4S2 Molecular Weight (g/mol): 240.292 InChI Key: LEVWYRKDKASIDU-IMJSIDKUSA-N Synonym: l-cystine,cystine,l-dicysteine,l-cystin,cystine acid,cysteine disulfide,oxidized l-cysteine,beta,beta'-dithiodialanine,h-cys-oh 2,l-cysteine disulfide PubChem CID: 67678 ChEBI: CHEBI:16283 IUPAC Name: (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid SMILES: C(C(C(=O)O)N)SSCC(C(=O)O)N
L-Cystine, specified according to the requirements of Ph.Eur.
CAS: 56-89-3 Molecular Formula: C6H12N2O4S2 Molecular Weight (g/mol): 240.292 InChI Key: LEVWYRKDKASIDU-IMJSIDKUSA-N Synonym: l-cystine,cystine,l-dicysteine,l-cystin,cystine acid,cysteine disulfide,oxidized l-cysteine,beta,beta'-dithiodialanine,h-cys-oh 2,l-cysteine disulfide PubChem CID: 67678 ChEBI: CHEBI:16283 IUPAC Name: (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid SMILES: C(C(C(=O)O)N)SSCC(C(=O)O)N