Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine disodium salt monohydrate, 98+%

Catalog No. AAJ6307118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6307118 50 g
AAJ6307130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6307118 Supplier Thermo Scientific Chemicals Supplier No. J6307118
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 199329-53-8 | C6H12N2Na2O5S2 | 302.271 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199329-53-8
Molecular Formula C6H12N2Na2O5S2
Molecular Weight (g/mol) 302.271
MDL Number MFCD00243291
InChI Key TVPDKJXPEMKMDM-BLZBDRIDSA-L
Synonym disodium l-cystine anion hydrate, l-cystine disodium salt monohydrate, h-cys-oh 2.2na.h2o, c6h10n2o4s2.2na.h2o
PubChem CID 71299670
IUPAC Name disodium;(2R)-2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate;hydrate
SMILES C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.O.[Na+].[Na+]

Specifications

Color White
Quantity 50 g
Formula Weight 302.27 (284.26 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material L-Cystine disodium salt monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 265-025-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.