Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine Disodium Salt, Monohydrate, Spectrum™ Chemical
SDP

Catalog No. 18620465 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-465 1 kg
18-620-463 25 g
18-620-464 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-465 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C30351KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cystine Sodium

Chemical Identifiers

CAS 199329-53-8

Specifications

Quantity 1 kg
Molecular Weight (g/mol) 302.26
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material L-Cystine Disodium Salt, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.