Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine EP/FCC, Ajinomoto

Catalog No. 502946093
Encompass
Change view
Click to view available options
Quantity:
1 kg
10 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-294-6093 10 kg
50-294-6045 1 kg
50-294-5990 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-294-6093 Supplier Thermo Scientific Production Chemicals and Services Supplier No. A34518
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56-89-3
Molecular Weight (g/mol) 240.3

Specifications

Packaging Poly Drum
Quantity 10 kg
Percent Purity 98.5 to 101.0%
Chemical Name or Material L-Cystine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.