Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine , MP Biomedicals, LLC

Catalog No. MP210145480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MP210145480 100 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210145480 Supplier MP Biomedicals, Inc Supplier No. 0210145480
Add to Cart
Add to Cart

L-Cystine is a crystalline solid with >99% purity.

  • Required for the synthesis of glutathione that protects the cells from oxidative stress
  • A specific x(c)-cysteine/glutamate antiporter mediates cellular uptake of cysteine in exchange for glutamate
  • Inhibition of cysteine uptake induces toxicity in neuronal cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.29
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, l-cysteine disulfide, --cystine
PubChem CID 67678
ChEBI CHEBI:16283

Specifications

Color White
pH 6
Melting Point >240°C (decomposition)(literature)
Quantity 100 g
Packaging Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.