Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Epicatechin, Mixt. of ca 60% L(-)-Epicatechin and ca 40% Catechin, Thermo Scientific™

Catalog No. AC226780010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226780010 1 g
AC226780050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226780010 Supplier Thermo Scientific Chemicals Supplier No. 226780010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-46-0
Molecular Formula C15H14O6
Molecular Weight (g/mol) 290.271
InChI Key PFTAWBLQPZVEMU-UKRRQHHQSA-N
Synonym --epicatechin, epicatechin, l-epicatechin, --epicatechol, l-acacatechin, epicatechol, l-epicatechol, 2r,3r-2-3,4-dihydroxyphenyl chroman-3,5,7-triol, -epicatechin, 2r,3r---epicatechin
PubChem CID 72276
ChEBI CHEBI:90
IUPAC Name (2R,3R)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)O

Specifications

Color Brown
Infrared Spectrum Authentic
Quantity 1 g
Beilstein 17, 214
Specific Rotation -31.5° to -33.5° (20°C, 589nm) (c=1, H2O:acetone 1:1)
Formula Weight 290.26
Percent Purity ∼60%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Epicatechin
EINECSNumber 207-710-1
RTECSNumber KB3745000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.