Learn More
Eurofins Discoverx L-Glutamic Acid, 10 mg. (92-1197)
Supplier:  Eurofins Discoverx 921197
Encompass
L-glutamic acid (glutamic acid) is a naturally produced proteinogenic non-essential amino acid that plays a role in neural activation. It is the most common excitatory neurotransmitter in the CNS. Acts as an agonist of the glutamate receptors (metabotropic, kainate, nMDA, and AMPA). May be dysregulated in nervous system disorders, such as epilepsy. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9725
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.