Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic Acid Monosodium Salt, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18611266 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-266 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-611-266 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GL135500GM

Chemical Identifiers

CAS 6106-04-3
Molecular Formula C5H10NNaO5
Molecular Weight (g/mol) 187.13
InChI Key GJBHGUUFMNITCI-QTNFYWBSSA-M
IUPAC Name sodium (4S)-4-amino-4-carboxybutanoate hydrate
SMILES O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O

Specifications

CAS Max % 1
Loss on Drying 0.005
Packaging Poly Bottle
Quantity 500 g
Specific Rotation +24.5° to +25.5°
Percent Purity 98.5 to 101.5%
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.