Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L(+)-Glutamic acid monosodium salt monohydrate, 99%

Catalog No. AC119940100
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119940100 10 kg Plastic Drum
AC119940010 1 kg Plastic Bottle
AC119940025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC119940100 Supplier Thermo Scientific Chemicals Supplier No. 119940100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 6106-04-03 | C5H8NNaO4·H2O | 187.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-04-03
Molecular Formula C5H8NNaO4·H2O
Molecular Weight (g/mol) 187.13
MDL Number MFCD00150138
InChI Key GJBHGUUFMNITCI-QTNFYWBSSA-M

Specifications

Melting Point 232.0°C
Color White
Chloride 2000ppm max
Infrared Spectrum Authentic
Assay Percent Range 0.99
Loss on Drying 0.5% max. (100°C, 5 hrs)
Packaging Plastic Drum
Linear Formula NaO2CCH2CH2CH(NH2)CO2H·H2O
Specific Rotation + 25.00
Sulfate 500ppm max
Synonym monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
Solubility Information Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol
SMILES C(CC(=O)O)C(C(=O)O)N.O.[Na]
IUPAC Name (2S)-2-aminopentanedioic acid;sodium;hydrate
Specific Rotation Condition + 25.00 (20.00°C c=10,2 N HCl)
Quantity 10 kg
PubChem CID 87090819
Formula Weight 187.13
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Glutamic acid monosodium salt monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.