Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic Acid Reagent, Reagents

Catalog No. 01337951
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-951 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-951 Supplier Reagents Holdings Llc Supplier No. C1210200100C2
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamic Acid Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Melting Point 213°C to 224°C
Boiling Point 175°C
pH 3.0 to 3.5
Color White to Off-white
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1.538
Grade Reagent
Physical Form Solid
Chemical Name or Material L-Glutamic Acid Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.