Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic Dehydrogenase, MP Biomedicals™

Catalog No. ICN15118980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50mg
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15118980 100mg
ICN15118950 50mg
ICN15118991 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15118980 Supplier MP Biomedicals, Inc Supplier No. 0215118980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 9029-12-3
Quantity 100mg
Synonym Glutamate dehydrogenase; L-glutamate dehydrogenase; L-glutamic dehydrogenase; Glutamic dehydrogenase; Glutamic acid dehydrogenase; GLDH; GDH; GluDH; L-GLDH; NAD(P)+-dependent glutamate dehydrogenase
Form Cloudy Suspension
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.