Learn More
Apexbio Technology LLC L-Glutamine(Synonyms: Glutamine, Levoglutamide, (S)-2-Aminoglutaramic acid, 2-Amino-4-carbamoylbutanoic acid), 100g, CAS: 56-85-9.
SDP
Supplier:  Apexbio Technology LLC A8461100000
Encompass_Preferred
L-Glutamine (CAS 56-85-9) is an amino acid abundantly expressed in human tissues and utilized extensively in cell metabolism particularly as a nitrogen and carbon donor Mechanistically L-glutamine serves as an essential substrate in glutaminolysis fueling the synthesis of nucleotides and glutathione and supporting energy production via the tricarboxylic acid (TCA) cycle Owing to its central role in cellular metabolism and proliferation L-glutamine is frequently investigated in research contexts involving gastrointestinal physiology immune cell function tissue repair processes and tumor cell metabolism
Catalog No. 50-000-77821
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.