Learn More
Quality Biological Inc L-Glutamine 200 mM, 100 mL
SDP
Supplier:  Quality Biological Inc 118084721EA
Encompass_Preferred
L-glutamine is a cell culture supplement, and an essential amnio acid. ¿ L-glutamine is required in cell culture for cellular growth. ¿ 100X concentration ¿ Applications: used in cell culture as an energy source in addition to carbohydrates (i.e. glucose, sucrose, maltose, etc.). ¿ L-glutamine participates in the formation of purine and pyrimidine nucleotides, amino sugars, gluthione, L-glutamate and other amino acids, as well as in protein synthesis and glucose production. ¿ Storage Conditions: -10¿C to -20¿C ¿ Shipping Condition: on dry ice ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 240 months
Catalog No. 50-751-7394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.