Learn More
Pfanstiehl Inc L-Glutamine, High Purity, Low Endotoxin, Low Metals, USP, JP, ChP (G-137) - 1KG, GMP, Multi-compendial - CAS: 56-85-9 - Injectable Excipient Grade
Supplier:  Pfanstiehl Inc G1371KG
Encompass
Pfanstiehl L-Glutamine, High Purity, Low Endotoxin, Low Metals is a multi-compendial amino acid that has been specifically designed and manufactured to remove impurities such a endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl L-Glutamine meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, vaccine, bioprocessing, and cell culture bioproduction feeds. Pfanstiehl L-Glutamine is available as USP (United States Pharmacopoeia), JP (Japanese Pharmacopoeia) and ChP (Chinese Pharmacopoeia). Compendial, U.S.P. , cGMP ,
Catalog No. NC2271932