Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-L-Glutamyl-p-nitroanilide Hydrochloride, MP Biomedicals™

Catalog No. ICN15149510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15149510 10 g
ICN15149501 1 g
ICN15149505 5 g
ICN15149525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15149510 Supplier MP Biomedicals, Inc Supplier No. 0215149510
Add to Cart
Add to Cart

Soluble in water (5mg/mL - clear, light yellow solution) or 0.1N HCl (1% w/v - clear, yellowish solution); soluble in methanol (100mg/mL with slight heat and constant stirring)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67953-08-6
Molecular Formula C11H14ClN3O5
Molecular Weight (g/mol) 303.70
MDL Number MFCD00039054
InChI Key WMRCHNNHOJNRSL-UHFFFAOYNA-N
Synonym gamma-l-glutamyl-p-nitroanilide, n-4-nitrophenyl-l-glutamine monohydrochloride, l-2-amino-4'-nitroglutaranilic acid hydrochloride, 2-amino-4-4-nitrophenyl carbamoyl butanoic acid hydrochloride, l-, a-glutamyl-p-nitroanilide, l-glutamic acid, a-p-nitroanilide hydrochloride, l-glutamic acid gamma-p-nitroanilide hydrochlori, l-glutamic acid gamma-p-nitroanilide hydrochloride, gamma-glutamyl transpeptidase substrate
IUPAC Name N-(3-carbamoyl-1-carboxypropyl)-4-nitroanilinium chloride
SMILES [Cl-].NC(=O)CCC([NH2+]C1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Color Beige
Quantity 10 g
Formula Weight 303.7
Physical Form Powder
Chemical Name or Material γ-L-Glutamyl-p-nitroanilide Hydrochloride
EINECSNumber 267-944-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.