Learn More
Cayman Chemical L-Glutathione, reduced, 70-18-8, 1 g
SDP
Supplier:  Cayman Chemical 100074611
Encompass_Preferred
Molecular Formula C10H17N3O6S, Purity ≥99%, Formula Weight 307.3, Synonyms: GSH; GSH is a tripeptide (-glutamylcysteinylglycine) widely distributed in both plants and animals.{1813,4672} GSH serves as a nucleophilic co-substrate to glutathione transferases in the detoxification of xenobiotics and is an essential electron donor to glutathione peroxidases in the reduction of hydroperoxides.{4672,165} GSH is also involved in amino acid transport and maintenance of protein sulfhydryl reduction status.{4669,4670} The concentration of GSH ranges from a few micromolar in plasma to several millimolar in tissues such as liver.
Catalog No. NC9418018
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.