Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-guanosine | 26578-09-6 | MFCD00064103 | 98.0% | C10H13N5O5 | 10MG
SDP

Supplier:  Medchemexpress LLC HY13813010MG

Encompass_Preferred

L-Guanosine is the L-enantiomer of the nucleoside guanosine (CAS 26578-09-6). It is supplied as a white to off-white solid intended for research use, exhibiting high purity and good solubility in DMSO; common applications include antiviral research and the preparation of supramolecular hydrogels.

  • Purity: 98.0%.
  • Chemical formula: C10H13N5O5.
  • Molecular weight: 283.24 g/mol.
  • Form: solid, white to off-white.
  • Solubility: soluble in DMSO (≈100 mg/mL); protocols available for aqueous formulations.
  • Storage: solid at 4°C protected from light; solutions at -80°C or -20°C with limited stability.
  • Intended use: research reagent for biochemical and materials science studies.

Catalog No. 50-002-60452


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.