Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(+)-Gulose 98%, Thermo Scientific™

Catalog No. AC257951000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC257951000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC257951000 Supplier Thermo Scientific Chemicals Supplier No. 257951000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6027-89-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00136022
InChI Key GZCGUPFRVQAUEE-JGWLITMVSA-N
Synonym l-gulose, aldehydo-l-gulose, 2s,3s,4r,5s-2,3,4,5,6-pentahydroxyhexanal, unii-j96e9q45n7
PubChem CID 80127
ChEBI CHEBI:37701
IUPAC Name (2S,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C=O

Specifications

Color Yellow
Infrared Spectrum Authentic
Quantity 100 mg
Merck Index 15, 4614
Specific Rotation +20° to +24° (20°C, 589nm) (c=5, H2O)
Formula Weight 180.16
Percent Purity ≥97.5%
Physical Form Viscous Oil
Chemical Name or Material L(+)-Gulose, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.