Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Gulose 98.0+%, TCI America™
SDP

Catalog No. G02391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02391G 1 g
G02395G 5 g
G023925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G02391G Supplier TCI America Supplier No. G02391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6027-89-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00136022
InChI Key GZCGUPFRVQAUEE-JGWLITMVSA-N
Synonym l-gulose, aldehydo-l-gulose, 2s,3s,4r,5s-2,3,4,5,6-pentahydroxyhexanal, unii-j96e9q45n7
PubChem CID 80127
ChEBI CHEBI:37701
IUPAC Name (2S,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C=O

Specifications

Melting Point 132°C
Color White
Quantity 1 g
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-Gulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.