Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidine Methyl Ester Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. H097725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H097725G 25 g
H09775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H097725G Supplier TCI America Supplier No. H097725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7389-87-9
Molecular Formula C7H11ClN3O2
Molecular Weight (g/mol) 204.63
MDL Number MFCD00012701
InChI Key VXXIPPPPKNWFLK-UHFFFAOYNA-N
Synonym l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
PubChem CID 2723645
IUPAC Name methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloridyl
SMILES [Cl].COC(=O)C(N)CC1=CN=CN1

Specifications

Color White
Quantity 25 g
Formula Weight 242.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine Methyl Ester Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.