Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Isoleucine, 99%, MP Biomedicals

Catalog No. 0219468901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-194-68901 1 g
ICN19468905 5 g
02-194-68925 25 g
02-194-68980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-194-68901 Supplier MP Biomedicals, Inc Supplier No. 0219468901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73-32-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064222 MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile
PubChem CID 6306
ChEBI CHEBI:17191
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N)C(O)=O

Specifications

Quantity 1 g
Formula Weight 131.2
Percent Purity 99.4%
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Isoleucine, 99%
Recommended Storage Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.