Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Isoleucine ethyl ester hydrochloride, 98%

Catalog No. AAH6464003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6464003 1 g
AAH6464006 5 g
AAH6464014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6464003 Supplier Thermo Scientific Chemicals Supplier No. H6464003

CAS: 56782-52-6 | C8H18ClNO2 | 195.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56782-52-6
Molecular Formula C8H18ClNO2
Molecular Weight (g/mol) 195.69
MDL Number MFCD09953729
InChI Key QQGRWNMNWONMOO-LEUCUCNGSA-N
Synonym l-isoleucine ethyl ester hydrochloride, h-ile-oet.hcl, ethyl l-isoleucinate hydrochloride, l-isoleucine ethyl ester hcl, 2s,3s-ethyl 2-amino-3-methylpentanoate hydrochloride, isoleucine, ethyl ester, hydrochloride, ethyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, l-isoleucine ethylester hcl, ksc497o6p, l-isoleucineethylesterhydrochloride
PubChem CID 6453390
SMILES Cl.CCOC(=O)[C@@H](N)[C@@H](C)CC

Specifications

Color White
Beilstein 4710859
IUPAC Name hydrogen ethyl (2S,3S)-2-amino-3-methylpentanoate chloride
Quantity 1 g
Formula Weight 195.69
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Isoleucine ethyl ester hydrochloride
EINECSNumber 260-382-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.