Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Kynurenine, R&D Systems™
SDP

Catalog No. 439350 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-935-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-935-0 Supplier R&D Systems Supplier No. 4393/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Tryptophan catabolite; endogenous aryl hydrocarbon receptor ligand

Tryptophan catabolite; endogenous activator of the aryl hydrocarbon receptor (AhR). Immunosuppressant; suppresses allogeneic T cell proliferation.

Chemical Identifiers

CAS 2922-83-0
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.22
MDL Number MFCD00069912
InChI Key YGPSJZOEDVAXAB-QMMMGPOBSA-N
Synonym l-kynurenine, s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, kynurenine, l, 3-anthraniloyl-l-alanine, unii-02jw4j5r44, ccris 4425, 2s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, quinurenine, kynurenin, s-alpha,2-diamino-gamma-oxobenzenebutanoic acid
PubChem CID 161166
ChEBI CHEBI:16946
IUPAC Name (2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid
SMILES N[C@@H](CC(=O)C1=CC=CC=C1N)C(O)=O

Specifications

Quantity 50 mg
Formula Weight 208.21
Percent Purity >98%
Product Line Tocris
Chemical Name or Material L-Kynurenine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.