Learn More
Abcam L-Leucine 7-Amido-4-Methylcoumarin, Fluorogenic Substrate, >98% Purity, 250 mg
Description
- Molecular weight: 288.37 g/mol
- Purity: >98%
- Application: Sensitive fluorogenic substrate for leucine aminopeptidase and aminopeptidase M
- Assay method: Hydrolysis rates can be measured using either a continuous or stopped assay
- Detection: The release of fluorescent 7-amino-4-methylcoumarin can be measured
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Substrate | L-Leucine 7-amido-4-methylcoumarin |
| Target Molecule | Leucine Aminopeptidase, Aminopeptidase M |
| Detection Method | Fluorescence |
| Quantity | 250 mg |
| For Use With (Application) | Leucine aminopeptidase and aminopeptidase M assays |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.