Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Leucine, 99%

Catalog No. AC125125000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC125125000 500 g
AC125121000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC125125000 Supplier Thermo Scientific Chemicals Supplier No. 125125000
  • $553.50
    $436.65 / Each of 1

    Save $116.85

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

L-Leucine, CAS # 61-90-5, is a branched-chain aliphatic essential amino acid in humans that is important for protein synthesis and many metabolic functions. | CAS: 61-90-5 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Leucine is an essential amino acid that has an important role in the growth and repair of muscle and bone tissue
  • It functions to preserve muscle glycogen and helps to prevent the breakdown of muscle proteins after trauma or severe stress
  • It is also involved in hemoglobin formation and metabolic functions. This amino acid cannot be synthesized to meet the body's needs from simpler molecules, and therefore must be provided in the diet.
  • L-Leucine stimulates protein synthesis by the initiation of mRNA translation

Applications

  • L-Leucine is used in the biosynthesis of proteins
  • In vivo studies show that L-Leucine can be used when gene defects lead to an organism's inability to metabolize the amino acid phenylalanine
  • This compound also contributes to the regulation of blood-sugar levels, growth hormone production, and wound healing
  • In research experiments this compound has been used as an activator of the mammalian target of rapamycin (mTOR)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Melting Point 286.0°C to 288.0°C
Specific Rotation + 15.40
Solubility Information Solubility in water: 22.4g/L (20°C). Other solubilities: 10.9g/L acetic acid, slightly soluble in alcohol, insoluble in ether
Sublimation Point 145.0 °C
Specific Rotation Condition + 15.40 (20.00°C c=4, 6N HCl)
Quantity 500 g
Formula Weight 131.17
Chemical Name or Material L-Leucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.