Learn More
U.S. Pharmacopeia L-Lysine Hydrochloride, 657-27-2, MFCD00064564, 200mg
Supplier: U.S. Pharmacopeia 1372005
Molecular formula C6H14N2O2 . HCl, Molecular weight 182.65, Melting Point 505.4 - 507.2 °F (263 - 264 °C), Synonyms: L-Lysine monohydrochloride
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.