Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lysine dihydrochloride, 99%

Catalog No. AAA1612714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1612714 25 g
AAA1612722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1612714 Supplier Thermo Scientific Chemicals Supplier No. A1612714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 657-26-1 | C6H16Cl2N2O2 | 219.11 g/mol

Synthesis of hyperbranched polylysine using the N-hydroxysuccinimide ester of L-lysine dihydrochloride as AB 2 building block. The histidine and lysine standards were prepared from L-histidine monohydro- chloride and L-lysine dihydrochloride respectively. L-Homocitrulline-The copper complex was prepared from L-lysine dihydrochloride in 80 to 86 per cent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of hyperbranched polylysine using the N-hydroxysuccinimide ester of L-lysine dihydrochloride as AB 2 building block. The histidine and lysine standards were prepared from L-histidine monohydro- chloride and L-lysine dihydrochloride respectively. L-Homocitrulline-The copper complex was prepared from L-lysine dihydrochloride in 80 to 86 per cent yields.

Solubility
Soluble in water (100 mg/ml).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-26-1
Molecular Formula C6H16Cl2N2O2
Molecular Weight (g/mol) 219.11
MDL Number MFCD00066389
InChI Key JBBURJFZIMRPCZ-XRIGFGBMSA-N
Synonym l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2
PubChem CID 69567
IUPAC Name (2S)-2,6-diaminohexanoic acid;dihydrochloride
SMILES Cl.Cl.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 202°C to 206°C
Beilstein 4539715
Merck Index 14,5636
Solubility Information Soluble in water (100mg/ml).
Quantity 25 g
Formula Weight 219.11
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Lysine dihydrochloride
Hazard Category H315-H320-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-518-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.