Learn More
L-(-)-Malic acid, 97%
Description
L-(-)-Malic acid is used as Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid rece. It also acts as active ingredient in many sour or tart foods. It is used as synthesizing disincrustant and fluorescent whitening agent. It aids in the production of polyester and alcohol acid resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-(-)-Malic acid is used as Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid rece. It also acts as active ingredient in many sour or tart foods. It is used as synthesizing disincrustant and fluorescent whitening agent. It aids in the production of polyester and alcohol acid resins.
Solubility
Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene
Notes
Incompatible with Bases, Oxidizing agents, Reducing agents, Alkali metals .
Specifications
Specifications
| Melting Point | 100°C to 106°C |
| Assay Percent Range | 97% |
| Beilstein | 1723541 |
| Merck Index | 14,5707 |
| Solubility Information | Soluble in acetone, dioxane, water, methanol and ethanol; Insoluble in benzene |
| Formula Weight | 134.09 |
| Percent Purity | 97% |
| Density | 1.6 |
| Quantity | 250 g |
| Flash Point | 220°C (428°F) |
| Show More |
Safety and Handling
| Hazard Category | H315-H318-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-601-5 |
| RTECSNumber | ON7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.