Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Methionine, [35S], In vitro Translation Grade , MP Biomedicals

Catalog No. MP0151001H5 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 μCi
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0151001H5 500 μCi
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0151001H5 Supplier MP Biomedicals, Inc Supplier No. 0151001H.5

  • Sp.Act. >1000 Ci/mmol; >37 TBq/mmol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH3S(CH2)2CH(NH2)COOH

Specifications

Physical Form Solution in 50 mM L-Lysine pH 7.4, containing 10 mM 2-mercaptoethanol
Quantity 500 μCi
Linear Formula CH3S(CH2)2CH(NH2)COOH
Formula Weight 149.2
Grade In vitro Translation
Chemical Name or Material L-METHIONINE, [35S], In vitro Translation Grade
Recommended Storage Store at -70°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.